Anti-AQP11 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5805BEA 348 EUR
ABGEAP5805B
Anti-AQP11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AQP11 (aquaporin 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR