Anti-FOXC2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5750CEA 350 EUR
ABGEAP5750C
Anti-FOXC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FOXC2 (forkhead box C2 (MFH-1, mesenchyme forkhead 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR