Anti-CACNA2D4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5708AEA 350 EUR
ABGEAP5708A
Anti-CACNA2D4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CACNA2D4 (calcium channel, voltage-dependent, alpha 2/delta subunit 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR