Anti-REG3G Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5606CEA 360 EUR
ABGEAP5606C
Anti-REG3G Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: REG3G (regenerating islet-derived 3 gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR