Anti-MC2R Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5540BEA 360 EUR
ABGEAP5540B
Anti-MC2R Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MC2R (melanocortin 2 receptor (adrenocorticotropic hormone))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR