Anti-DDC Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5536AEA 350 EUR
ABGEAP5536A
Anti-DDC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDC (dopa decarboxylase (aromatic L-amino acid decarboxylase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR