Anti-RPS3A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5529BEA 350 EUR
ABGEAP5529B
Anti-RPS3A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPS3A (ribosomal protein S3A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR