Anti-EHMT2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5526AEA 348 EUR
ABGEAP5526A
Anti-EHMT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EHMT2 (euchromatic histone-lysine N-methyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR