Anti-ITGA5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5519BEA 360 EUR
ABGEAP5519B
Anti-ITGA5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITGA5 (integrin, alpha 5 (fibronectin receptor, alpha polypeptide))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR