Anti-MEIS2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5518CEA 348 EUR
ABGEAP5518C
Anti-MEIS2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MEIS2 (Meis homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR