Anti-USP17L2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5491BEA 348 EUR
ABGEAP5491B
Anti-USP17L2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: USP17L2 (ubiquitin specific peptidase 17-like family member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR