Anti-DKC1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5479CEA 350 EUR
ABGEAP5479C
Anti-DKC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DKC1 (dyskeratosis congenita 1, dyskerin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR