Anti-DYNC1LI2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5478CEA 350 EUR
ABGEAP5478C
Anti-DYNC1LI2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DYNC1LI2 (dynein, cytoplasmic 1, light intermediate chain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR