Anti-TFAP4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5441BEA 350 EUR
ABGEAP5441B
Anti-TFAP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TFAP4 (transcription factor AP-4 (activating enhancer binding protein 4))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR