Anti-SMIM14 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5437BEA 348 EUR
ABGEAP5437B
Anti-SMIM14 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMIM14 (small integral membrane protein 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR