Anti-DEPDC1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5428AEA 360 EUR
ABGEAP5428A
Anti-DEPDC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DEPDC1 (DEP domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR