Anti-IFT43 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5370CEA 350 EUR
ABGEAP5370C
Anti-IFT43 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IFT43 (intraflagellar transport 43 homolog (Chlamydomonas))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR