Anti-ATP6V0A4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5369CEA 350 EUR
ABGEAP5369C
Anti-ATP6V0A4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATP6V0A4 (ATPase, H+ transporting, lysosomal V0 subunit a4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR