Anti-WDR18 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5355BEA 350 EUR
ABGEAP5355B
Anti-WDR18 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: WDR18 (WD repeat domain 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR