Anti-UTP6 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5296AEA 348 EUR
ABGEAP5296A
Anti-UTP6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UTP6 (UTP6, small subunit (SSU) processome component, homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR