Anti-HAUS5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5230AEA 350 EUR
ABGEAP5230A
Anti-HAUS5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HAUS5 (HAUS augmin-like complex, subunit 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR