Anti-ENTPD8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5161BEA 350 EUR
ABGEAP5161B
Anti-ENTPD8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ENTPD8 (ectonucleoside triphosphate diphosphohydrolase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR