Anti-G6PD Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5094CEA 348 EUR
ABGEAP5094C
Anti-G6PD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: G6PD (glucose-6-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR