Anti-TPRGL Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5083AEA 348 EUR
ABGEAP5083A
Anti-TPRGL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TPRGL
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR