Anti-SGTB Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5076AEA 360 EUR
ABGEAP5076A
Anti-SGTB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SGTB (small glutamine-rich tetratricopeptide repeat (TPR)-containing, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR