Anti-NAA50 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5060BEA 350 EUR
ABGEAP5060B
Anti-NAA50 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NAA50 (N(alpha)-acetyltransferase 50, NatE catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR