Anti-FOLR2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5032AEA 360 EUR
ABGEAP5032A
Anti-FOLR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FOLR2 (folate receptor 2 (fetal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR