Anti-EFR3A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4982BEA 350 EUR
ABGEAP4982B
Anti-EFR3A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EFR3A (EFR3 homolog A (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR