Anti-PAN3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4958BEA 350 EUR
ABGEAP4958B
Anti-PAN3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAN3 (PAN3 poly(A) specific ribonuclease subunit homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR