Anti-KCNG3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4954AEA 348 EUR
ABGEAP4954A
Anti-KCNG3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KCNG3 (potassium voltage-gated channel, subfamily G, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR