Anti-POLR3H Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4936AEA 350 EUR
ABGEAP4936A
Anti-POLR3H Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POLR3H (polymerase (RNA) III (DNA directed) polypeptide H (22.9kD))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR