Anti-PLA2G1B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4878BEA 350 EUR
ABGEAP4878B
Anti-PLA2G1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLA2G1B (phospholipase A2, group IB (pancreas))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR