Anti-PRPF19 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4870AEA 350 EUR
ABGEAP4870A
Anti-PRPF19 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRPF19 (pre-mRNA processing factor 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR