Anti-CCR1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4859AEA 350 EUR
ABGEAP4859A
Anti-CCR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCR1 (chemokine (C-C motif) receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR