Anti-GABRG3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4800AEA 350 EUR
ABGEAP4800A
Anti-GABRG3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GABRG3 (gamma-aminobutyric acid (GABA) A receptor, gamma 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR