Anti-DDO Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4795CEA 348 EUR
ABGEAP4795C
Anti-DDO Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDO (D-aspartate oxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR