Anti-HIF1A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4776AEA 360 EUR
ABGEAP4776A
Anti-HIF1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR