Anti-TOR1B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4771CEA 348 EUR
ABGEAP4771C
Anti-TOR1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TOR1B (torsin family 1, member B (torsin B))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR