Anti-RDH10 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4738CEA 360 EUR
ABGEAP4738C
Anti-RDH10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RDH10 (retinol dehydrogenase 10 (all-trans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR