Anti-GL8D2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4719AEA 350 EUR
ABGEAP4719A
Anti-GL8D2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GL8D2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR