Anti-ATL3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP4715CEA 348 EUR
ABGEAP4715C
Anti-ATL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATL3 (atlastin GTPase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR