Anti-GATA6 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3766AEA 360 EUR
ABGEAP3766A
Anti-GATA6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GATA6 (GATA binding protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR