Anti-NPHS1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3747AEA 348 EUR
ABGEAP3747A
Anti-NPHS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NPHS1 (nephrosis 1, congenital, Finnish type (nephrin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR