Anti-MAP1LC3B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3744AEA 360 EUR
ABGEAP3744A
Anti-MAP1LC3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP1LC3B (microtubule-associated protein 1 light chain 3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR