Anti-LINGO-1(LRRN6A) Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3685AEA 360 EUR
ABGEAP3685A
Anti-LINGO-1(LRRN6A) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LINGO-1(LRRN6A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR