Anti-NOS3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3665AEA 370 EUR
ABGEAP3665A
Anti-NOS3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NOS3 (nitric oxide synthase 3 (endothelial cell))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR