Anti-IL6ST Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3663AEA 360 EUR
ABGEAP3663A
Anti-IL6ST Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL6ST (interleukin 6 signal transducer (gp130, oncostatin M receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR