Anti-MUC1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3614BEA 360 EUR
ABGEAP3614B
Anti-MUC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR