Anti-PRL Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3573AEA 360 EUR
ABGEAP3573A
Anti-PRL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRL (prolactin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR