Anti-DNMT1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3534AEA 360 EUR
ABGEAP3534A
Anti-DNMT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNMT1 (DNA (cytosine-5-)-methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR